accessibility menu, dialog, popup

UserName

H3C(S)OH

CAS RN 1445-91-6

IUPAC Name: (1S)-1-phenylethan-1-ol
Synonyms: (S)-1-phenylethanol (S)-α-methylbenzenemethanol (S)-1-phenethyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: WAPNOHKVXSQRPX-ZETCQYMHSA-N
SMILES: C[C@H](O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)