missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| (S,S)-Ts-DENEB(regR) | |
| C31H33ClN2O3RuS | |
| 1 g | |
| Chloro[(S,S)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(S,S)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) | |
| CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+] | |
| 650.196 | |
| 650.20 |
| 1384974-37-1 | |
| MFCD22666040 | |
| 3077 | |
| INKUCOHLIHBSDN-YBZGWEFGSA-M | |
| chlororuthenium(1+);[(1S,2S)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide | |
| 121235453 | |
| Crystalline Powder |
Chemical Identifiers
| 1384974-37-1 | |
| 650.196 | |
| INKUCOHLIHBSDN-YBZGWEFGSA-M | |
| 121235453 | |
| CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+] |
| C31H33ClN2O3RuS | |
| MFCD22666040 | |
| Chloro[(S,S)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(S,S)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) | |
| chlororuthenium(1+);[(1S,2S)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide |
Safety and Handling
TSCA : No