missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-Ts-DENEB(regR), TCI America™

Catalog No. T3077200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3077200MG 200 mg
T30771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3077200MG Supplier TCI America Supplier No. T3077200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1384974-37-1
Molecular Formula C31H33ClN2O3RuS
Molecular Weight (g/mol) 650.196
MDL Number MFCD22666040
InChI Key INKUCOHLIHBSDN-YBZGWEFGSA-M
Synonym Chloro[(S,S)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(S,S)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 121235453
IUPAC Name chlororuthenium(1+);[(1S,2S)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+]

Specifications

Quantity 200 mg
UN Number 3077
Formula Weight 650.20
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-Ts-DENEB(regR)