missing translation for 'onlineSavingsMsg'
Learn More

(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine 98.0+%, TCI America™

Catalog No. p-7128594
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4995200MG 200 mg
D49951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4995200MG Supplier TCI America Supplier No. D4995200MG

Chemical Identifiers

CAS 209482-27-9
Molecular Formula C36H30NO2P
Molecular Weight (g/mol) 539.62
MDL Number MFCD04117688,MFCD08561138
InChI Key LKZPDRCMCSBQFN-UHFFFAOYNA-N
Synonym (11bS)-N,N-Bis[(1S)-1-phenylethyl]dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine
PubChem CID 23592741
IUPAC Name N,N-bis(1-phenylethyl)-12,14-dioxa-13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-amine
SMILES CC(N(C(C)C1=CC=CC=C1)P1OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 539.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine