accessibility menu, dialog, popup

UserName

H3CNCH3POO

CAS RN 209482-27-9

IUPAC Name: N,N-bis(1-phenylethyl)-12,14-dioxa-13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-amine
Synonyms: N,N-bis(1-phenylethyl)-12,14-dioxa-13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-amine
Molecular Weight (g/mol): 539.62
Molecular Formula: C36H30NO2P
InChi Key: LKZPDRCMCSBQFN-UHFFFAOYNA-N
SMILES: CC(N(C(C)C1=CC=CC=C1)P1OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results