missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America E101125G
Specifications
| (S,S)-N,N′-Bis(1-hydroxy-2-butyl)ethylenediamine Dihydrochloride | |
| 199°C | |
| C10H26Cl2N2O2 | |
| 25 g | |
| AUAHHJJRFHRVPV-HSTMFJOWSA-N | |
| (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride | |
| 6419929 | |
| ≥98.0% (T) |
| 1070-11-7 | |
| White | |
| MFCD00216025 | |
| ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride | |
| CCC(CO)NCCNC(CC)CO.Cl.Cl | |
| 277.23 | |
| 277.23 | |
| Crystalline Powder |
Chemical Identifiers
| 1070-11-7 | |
| 277.23 | |
| AUAHHJJRFHRVPV-HSTMFJOWSA-N | |
| 6419929 | |
| CCC(CO)NCCNC(CC)CO.Cl.Cl |
| C10H26Cl2N2O2 | |
| MFCD00216025 | |
| ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride | |
| (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride |
Safety and Handling
RTECSNumber : EL3854000
TSCA : No