missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7129278
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10115G 5 g
E101125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10115G Supplier TCI America Supplier No. E10115G
Only null left

Chemical Identifiers

CAS 1070-11-7
Molecular Formula C10H26Cl2N2O2
Molecular Weight (g/mol) 277.23
MDL Number MFCD00216025
InChI Key AUAHHJJRFHRVPV-HSTMFJOWSA-N
Synonym ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride
PubChem CID 6419929
IUPAC Name (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride
SMILES CCC(CO)NCCNC(CC)CO.Cl.Cl

Specifications

Melting Point 199°C
Color White
Quantity 5 g
Formula Weight 277.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-N,N′-Bis(1-hydroxy-2-butyl)ethylenediamine Dihydrochloride
RTECSNumber EL3854000
TSCA No