missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-Hydrobenzoin, 98+%

Catalog No. p-7058011
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1408206 5 g
AAL1408214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1408206 Supplier Thermo Scientific Chemicals Supplier No. L1408206
Only null left

CAS: 2325-10-2 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2325-10-2
MDL Number MFCD00064255

Specifications

Melting Point 148°C to 150°C
Color White
Molecular Formula C11H16ClNO2
Quantity 5 g
Beilstein 2330888
Merck Index 14,4777
Synonym 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N
SMILES COC1=C(C=C2CNCCC2=C1)OC.Cl
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
Molecular Weight (g/mol) 229.70
PubChem CID 6456035
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material (S,S)-(-)-Hydrobenzoin
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only