missing translation for 'onlineSavingsMsg'
Learn More

S,S-Diphenylsulfilimine Monohydrate 98.0+%, TCI America™

Catalog No. D20025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20025G 5 g
D20021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20025G Supplier TCI America Supplier No. D20025G
Only null left

Chemical Identifiers

CAS 68837-61-6
Molecular Formula C12H13NOS
Molecular Weight (g/mol) 219.302
MDL Number MFCD00001757
InChI Key YLGYIQQZVOCMDO-UHFFFAOYSA-N
PubChem CID 11435938
IUPAC Name imino(diphenyl)-$l^{4}-sulfane;hydrate
SMILES C1=CC=C(C=C1)S(=N)C2=CC=CC=C2.O

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S,S-Diphenylsulfilimine Monohydrate