accessibility menu, dialog, popup

UserName

SHNH2O

CAS RN 68837-61-6

IUPAC Name: (diphenyl-λ⁴-sulfanylidene)amine hydrate
Synonyms: (diphenyl-λ⁴-sulfanylidene)amine hydrate
Molecular Weight (g/mol): 219.3
Molecular Formula: C12H13NOS
InChi Key: YLGYIQQZVOCMDO-UHFFFAOYSA-N
SMILES: O.N=S(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)