missing translation for 'onlineSavingsMsg'
Learn More

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%

CAS: 22236-45-9 | C9H13NO3S2 | 247.327 g/mol

$70.26 - $195.95

Chemical Identifiers

CAS 22236-45-9
Molecular Formula C9H13NO3S2
Molecular Weight (g/mol) 247.327
MDL Number MFCD00013922
InChI Key IRNAWARRPQUZDU-UHFFFAOYSA-N
Synonym s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
PubChem CID 99226
IUPAC Name N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0057706
View Documents
Thermo Scientific Chemicals
L0057706
5 g N/A
Only null left
 
AAL0057714
View Documents
Thermo Scientific Chemicals
L0057714
25 g N/A
Only null left
 
Description

Description

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

22236-45-9
247.327
IRNAWARRPQUZDU-UHFFFAOYSA-N
99226
CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C
C9H13NO3S2
MFCD00013922
s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide

Specifications

22236-45-9
C9H13NO3S2
5 g
s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
IRNAWARRPQUZDU-UHFFFAOYSA-N
N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide
99226
98%
167°C to 169°C
MFCD00013922
2119329
Soluble in EtOH,THF,DMSO.
CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C
247.327
247.34
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 244-861-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only