missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-Rolipram 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 134°C |
| Color | White-Yellow |
| Quantity | 10 mg |
| Formula Weight | 275.35 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (S)-(+)-Rolipram |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |