missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Propylene Oxide 98.0+%, TCI America™
$34.66 - $34.66
Chemical Identifiers
| CAS | 16088-62-3 |
|---|---|
| Molecular Formula | C3H6O |
| Molecular Weight (g/mol) | 58.08 |
| MDL Number | MFCD00064312 |
| InChI Key | GOOHAUXETOMSMM-VKHMYHEASA-N |
| Synonym | s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane |
| PubChem CID | 146262 |
| ChEBI | CHEBI:28982 |
| IUPAC Name | (2S)-2-methyloxirane |
| SMILES | C[C@H]1CO1 |
Chemical Identifiers
| 16088-62-3 | |
| 58.08 | |
| GOOHAUXETOMSMM-VKHMYHEASA-N | |
| 146262 | |
| (2S)-2-methyloxirane |
| C3H6O | |
| MFCD00064312 | |
| s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane | |
| CHEBI:28982 | |
| C[C@H]1CO1 |
Specifications
| 16088-62-3 | |
| 34°C | |
| MFCD00064312 | |
| 1280 | |
| GOOHAUXETOMSMM-VKHMYHEASA-N | |
| (2S)-2-methyloxirane | |
| 146262 | |
| 58.08 | |
| Liquid |
| Colorless | |
| C3H6O | |
| 1 mL | |
| s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane | |
| C[C@H]1CO1 | |
| 58.08 | |
| CHEBI:28982 | |
| ≥98.0% (GC) | |
| (S)-(-)-Propylene Oxide |
Safety and Handling
EINECSNumber : (2)-0219
RTECSNumber : UJ2651000
TSCA : No
Recommended Storage : Refrigerator