accessibility menu, dialog, popup

UserName

CAS RN 16088-62-3

IUPAC Name: (2S)-2-methyloxirane
Synonyms: (-)-propylene oxide S-methyloxirane (S)-epoxypropane S-epoxypropane (-)-methyloxirane
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: GOOHAUXETOMSMM-VKHMYHEASA-N
SMILES: C[C@H]1CO1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)