missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Propylene Oxide 98.0+%, TCI America™

Catalog No. P09511ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09511ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09511ML Supplier TCI America Supplier No. P09511ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16088-62-3
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00064312
InChI Key GOOHAUXETOMSMM-VKHMYHEASA-N
Synonym s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane
PubChem CID 146262
ChEBI CHEBI:28982
IUPAC Name (2S)-2-methyloxirane
SMILES C[C@H]1CO1

Specifications

Color Colorless
Boiling Point 34°C
Quantity 1 mL
UN Number 1280
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-Propylene Oxide
EINECSNumber (2)-0219
RTECSNumber UJ2651000
TSCA No
Recommended Storage Refrigerator