missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Propylene oxide, 99%

Catalog No. p-7026373
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2234106 5 g
AAB2234114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2234106 Supplier Thermo Scientific Chemicals Supplier No. B2234106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16088-62-3 | C3H6O | 58.08 g/mol

(S)-(-)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used in making the polyurethane plastics. It finds an application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It is also used in model aircraft and surface vehicles as a glow fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used in making the polyurethane plastics. It finds an application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It is also used in model aircraft and surface vehicles as a glow fuel.

Solubility
Miscible with water and many organic solvents.

Notes
Highly flammable. Keep away from sources of ignition. The substance may polymerize under the influence of bases, acids and metal chlorides.

Chemical Identifiers

CAS 16088-62-3
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00064312
InChI Key GOOHAUXETOMSMM-VKHMYHEASA-N
Synonym s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane
PubChem CID 146262
ChEBI CHEBI:28982
IUPAC Name (2S)-2-methyloxirane
SMILES C[C@H]1CO1

Specifications

Density 0.829
Boiling Point 34°C to 35°C
Flash Point −37°C (−35°F)
Odor Ether-like
Refractive Index 1.367
Quantity 5 g
UN Number UN1280
Beilstein 79765
Merck Index 14,7856
Solubility Information Miscible with water and many organic solvents.
Optical Rotation −14° (Neat)
Formula Weight 58.08
Percent Purity 99%
Chemical Name or Material (S)-(-)-Propylene oxide
Show More Show Less
Hazard Category H224-H302-H311+H331-H319-H335-H340-H350
Hazard Statement GHS H Statement
H224-H340-H350-H302-H312-H332-H315-H319-H335
Extremely flammable liquid and vapour.
May cause genetic defects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: PROPYLENE OXIDE
EINECSNumber 240-241-0
RTECSNumber UJ2651000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only