missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Potassium Thioacetate 97.0+%, TCI America™
Supplier: TCI America T203025G
Specifications
| S-Potassium Thioacetate | |
| 176°C | |
| C2H4KOS | |
| 25 g | |
| SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
| ethanethioic S-acid;potassium | |
| 24201352 | |
| ≥97.0% (T) |
| 10387-40-3 | |
| White-Yellow | |
| MFCD00083065 | |
| potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
| CC(=O)S.[K] | |
| 115.211 | |
| 114.20 | |
| Crystalline Powder |
Chemical Identifiers
| 10387-40-3 | |
| 115.211 | |
| SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
| 24201352 | |
| CC(=O)S.[K] |
| C2H4KOS | |
| MFCD00083065 | |
| potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
| ethanethioic S-acid;potassium |
Safety and Handling
EINECSNumber : (2)-3702
TSCA : Yes