missing translation for 'onlineSavingsMsg'
Learn More

S-Potassium Thioacetate 97.0+%, TCI America™

Catalog No. p-7137069
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T203025G 25 g
T2030250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T203025G Supplier TCI America Supplier No. T203025G
Only null left

Chemical Identifiers

CAS 10387-40-3
Molecular Formula C2H4KOS
Molecular Weight (g/mol) 115.211
MDL Number MFCD00083065
InChI Key SDJHDRMYZQFJJO-UHFFFAOYSA-N
Synonym potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
PubChem CID 24201352
IUPAC Name ethanethioic S-acid;potassium
SMILES CC(=O)S.[K]

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
Formula Weight 114.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-Potassium Thioacetate
EINECSNumber (2)-3702
TSCA Yes