missing translation for 'onlineSavingsMsg'
Learn More

S-Potassium Thioacetate 97.0+%, TCI America™

Supplier:  TCI America T2030250G

 View more versions of this product

Catalog No. T2030250G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

S-Potassium Thioacetate
176°C
C2H4KOS
250 g
SDJHDRMYZQFJJO-UHFFFAOYSA-N
ethanethioic S-acid;potassium
24201352
≥97.0% (T)
10387-40-3
White-Yellow
MFCD00083065
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
CC(=O)S.[K]
115.211
114.20
Crystalline Powder

Chemical Identifiers

10387-40-3
115.211
SDJHDRMYZQFJJO-UHFFFAOYSA-N
24201352
CC(=O)S.[K]
C2H4KOS
MFCD00083065
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
ethanethioic S-acid;potassium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3702

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions