missing translation for 'onlineSavingsMsg'
Learn More

S-Phenyl thioacetate, 98%

Catalog No. p-7052655
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5517506 5 g
AAH5517514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5517506 Supplier Thermo Scientific Chemicals Supplier No. H5517506

CAS: 934-87-2 | C8H8OS | 152.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 934-87-2
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00008752
InChI Key WBISVCLTLBMTDS-UHFFFAOYSA-N
Synonym s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl
PubChem CID 13630
IUPAC Name S-phenyl ethanethioate
SMILES CC(=O)SC1=CC=CC=C1

Specifications

Color Colorless
Density 1.124 g/mL
Boiling Point 99°C to 100°C (6 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.57
Linear Formula CH3COSC6H5
Quantity 5 g
Formula Weight 152.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material S-Phenyl thioacetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-294-2
RTECSNumber AJ7559000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only