missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-N-BOC-3-pyrrolidinol, 98+%, Thermo Scientific™

Chemical Identifiers

CAS 101469-92-5
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 178.24
MDL Number MFCD01317839
InChI Key APCBTRDHCDOPNY-ZETCQYMHSA-N
Synonym s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
PubChem CID 854055
IUPAC Name tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397580050
SDS View Documents
Thermo Scientific Chemicals
397580050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

101469-92-5
178.24
APCBTRDHCDOPNY-ZETCQYMHSA-N
854055
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD01317839
s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate

Specifications

101469-92-5
100.0
Authentic
Glass bottle
MFCD01317839
+21° to +26° (20°C, 589nm) (C=1, in Methanol)
APCBTRDHCDOPNY-ZETCQYMHSA-N
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
854055
98+%
98.0
273.3°C
98% min. (HPLC)
C9H17NO3
5g
s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
CC(C)(C)OC(=O)N1CCC(C1)O
178.24
178.24
(S)-(+)-N-BOC-3-pyrrolidinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only