accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHO

CAS RN 101469-92-5

IUPAC Name: tert-butyl 3-hydroxypyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-hydroxypyrrolidine-1-carboxylate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: APCBTRDHCDOPNY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(O)C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)