missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-α-Methylbenzyl isocyanate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(S)-(-)-alpha-Methylbenzyl isocyanate is a useful chiral derivatization reagent for all gas chromatography (GC) applications in the chiral field. It has been employed to meet the requirements for derivatization reagents for enantiomeric excess determinations.
$334.01 - $1270.64
Chemical Identifiers
| MDL Number | MFCD00002036 |
|---|---|
| Synonym | (S)-(-)-1-Phenylethyl isocyanate |
Chemical Identifiers
| MFCD00002036 | |
| (S)-(-)-1-Phenylethyl isocyanate |
Specifications
| 14649-03-7 | |
| 55°C to 56°C/2.5 mmHg | |
| C6H5 CH(CH3)NCO | |
| 1 mL | |
| 147.17 | |
| For chiral derivatization |
| 1.045 g/mL (at 20°C) | |
| 65°C - Closed Cup | |
| MFCD00002036 | |
| (S)-(-)-1-Phenylethyl isocyanate | |
| ≥99.0% (sum of enantiomers, GC); ≥99.0% |