missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-α-Methylbenzyl isocyanate, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018760
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018760 1 mL
111018029 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018760 Supplier Merck Emd Millipore Supplier No. 779701ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(S)-(-)-alpha-Methylbenzyl isocyanate is a useful chiral derivatization reagent for all gas chromatography (GC) applications in the chiral field. It has been employed to meet the requirements for derivatization reagents for enantiomeric excess determinations.

Chemical Identifiers

MDL Number MFCD00002036
Synonym (S)-(-)-1-Phenylethyl isocyanate

Specifications

Density 1.045 g/mL (at 20°C)
CAS 14649-03-7
Boiling Point 55°C to 56°C/2.5 mmHg
Flash Point 65°C - Closed Cup
Linear Formula C6H5 CH(CH3)NCO
Quantity 1 mL
Formula Weight 147.17
Percent Purity ≥99.0% (sum of enantiomers, GC); ≥99.0%
Grade For chiral derivatization