missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Limonene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. p-7187504
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020203 1 mL
111018729 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020203 Supplier Merck Emd Millipore Supplier No. 621281ML
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 5989-54-8
Vapor Pressure <3 mmHg (14.4°C)
Vapor Density 4.7 (vs. air)
Density 0.844 g/mL (at 25°C)
Flash Point 51°C - closed cup
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00001558
Boiling Point 175°C to 177°C
Synonym (-)-p-Mentha-1,8-diene; (-)-Carvene; (S)-4-Isopropenyl-1-methyl cyclohexene
Type Analytical Standard
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less