Learn More
(S)-(-)-Limonene, 97%
CAS: 5989-54-8 | C10H16 | 136.238 g/mol
$69.01 - $167.53
Chemical Identifiers
| CAS | 5989-54-8 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.238 |
| MDL Number | MFCD00001558 |
| InChI Key | XMGQYMWWDOXHJM-SNVBAGLBSA-N |
| Synonym | --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s-- |
| PubChem CID | 439250 |
| ChEBI | CHEBI:15383 |
| IUPAC Name | (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene |
| SMILES | CC1=CCC(CC1)C(=C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1324418
|
Thermo Scientific Chemicals
L1324418 |
50 g |
Each for $69.01
|
|
|||||
|
AAL1324430
|
Thermo Scientific Chemicals
L1324430 |
250 g |
Each for $167.53
|
|
|||||
Description
(S)-(-)-Limonene is used to inhibit the proliferation of colon cancer cells. It is also used in artificial essential oils in order to produce (+)-carvone. It is an active component of turpentine. Further, it is used in flavorings, fragrances, and cosmetics. It finds application as a solvent and wetting agent. In addition to this it is used to produce resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-Limonene is used to inhibit the proliferation of colon cancer cells. It is also used in artificial essential oils in order to produce (+)-carvone. It is an active component of turpentine. Further, it is used in flavorings, fragrances, and cosmetics. It finds application as a solvent and wetting agent. In addition to this it is used to produce resins.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible strong oxidizing agents, acids, strong bases and peroxides.
Chemical Identifiers
| 5989-54-8 | |
| 136.238 | |
| XMGQYMWWDOXHJM-SNVBAGLBSA-N | |
| 439250 | |
| (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene |
| C10H16 | |
| MFCD00001558 | |
| --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s-- | |
| CHEBI:15383 | |
| CC1=CCC(CC1)C(=C)C |
Specifications
| 5989-54-8 | |
| 0.844 | |
| 48°C (118°F) | |
| 1.472 | |
| 50 g | |
| 2323991 | |
| --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s-- | |
| XMGQYMWWDOXHJM-SNVBAGLBSA-N | |
| CC1=CCC(CC1)C(=C)C | |
| 136.238 | |
| CHEBI:15383 | |
| 97% |
| -74°C | |
| 175°C to 177°C | |
| C10H16 | |
| MFCD00001558 | |
| UN2052 | |
| 14,5493 | |
| Miscible with alcohol. Immiscible with water. | |
| −90° (c=10 in Ethanol) | |
| (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene | |
| 439250 | |
| 136.23 | |
| (S)-(-)-Limonene |
Safety and Handling
GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c
H226-H304-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE
EINECSNumber : 227-815-6
RTECSNumber : OS8350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only