missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Limonene, 97%

CAS: 5989-54-8 | C10H16 | 136.238 g/mol

Supplier:  Thermo Scientific Chemicals L1324430

 View more versions of this product

Catalog No. AAL1324430


Only null left

Description

Description

(S)-(-)-Limonene is used to inhibit the proliferation of colon cancer cells. It is also used in artificial essential oils in order to produce (+)-carvone. It is an active component of turpentine. Further, it is used in flavorings, fragrances, and cosmetics. It finds application as a solvent and wetting agent. In addition to this it is used to produce resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-Limonene is used to inhibit the proliferation of colon cancer cells. It is also used in artificial essential oils in order to produce (+)-carvone. It is an active component of turpentine. Further, it is used in flavorings, fragrances, and cosmetics. It finds application as a solvent and wetting agent. In addition to this it is used to produce resins.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible strong oxidizing agents, acids, strong bases and peroxides.
Specifications

Specifications

(S)-(-)-Limonene
-74°C
175°C to 177°C
C10H16
MFCD00001558
UN2052
14,5493
Miscible with alcohol. Immiscible with water.
−90° (c=10 in Ethanol)
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
439250
136.23
5989-54-8
0.844
48°C (118°F)
1.472
250 g
2323991
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
XMGQYMWWDOXHJM-SNVBAGLBSA-N
CC1=CCC(CC1)C(=C)C
136.238
CHEBI:15383
97%

Chemical Identifiers

5989-54-8
136.238
XMGQYMWWDOXHJM-SNVBAGLBSA-N
439250
(4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
C10H16
MFCD00001558
--limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
CHEBI:15383
CC1=CCC(CC1)C(=C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.

P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c

H226-H304-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE

EINECSNumber : 227-815-6

RTECSNumber : OS8350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only