missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ketopinic Acid 98.0+%, TCI America™

Catalog No. K00285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00285G 5 g
K00281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00285G Supplier TCI America Supplier No. K00285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40724-67-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD08061242
InChI Key WDODWBQJVMBHCO-UHFFFAOYNA-N
PubChem CID 739183
IUPAC Name 7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-carboxylic acid
SMILES CC1(C)C2CCC1(C(O)=O)C(=O)C2

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-Ketopinic Acid