accessibility menu, dialog, popup

UserName

CAS RN 40724-67-2

IUPAC Name: 7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-carboxylic acid
Synonyms: 7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-carboxylic acid
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: WDODWBQJVMBHCO-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C(O)=O)C(=O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results