missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Glycidyl Phenyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 1 g |
| Formula Weight | 150.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-Glycidyl Phenyl Ether |
Safety and Handling
| EINECSNumber | (3)-0559&(3)-0594 |
| TSCA | No |