CAS RN 71031-03-3
IUPAC Name:
2-(phenoxymethyl)oxirane
Synonyms:
phenyl glydidyl ether
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
FQYUMYWMJTYZTK-UHFFFAOYNA-N
SMILES:
C(OC1=CC=CC=C1)C1CO1
1
–
1
of
1
results
1
(S)-Glycidyl Phenyl Ether 98.0+%, TCI America™
CAS: 71031-03-3 Molecular Formula: C9H10O2 Molecular Weight (g/mol): 150.177 MDL Number: MFCD06659026 InChI Key: FQYUMYWMJTYZTK-SECBINFHSA-N Synonym: (S)-2-(Phenoxymethyl)oxirane, (S)-1,2-Epoxy-3-phenoxypropane PubChem CID: 1534345 IUPAC Name: (2S)-2-(phenoxymethyl)oxirane SMILES: C1C(O1)COC2=CC=CC=C2