missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl Butyrate 98.0+%, TCI America™

Catalog No. G02835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02835G 5 g
G028325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02835G Supplier TCI America Supplier No. G02835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65031-96-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00273369
InChI Key YLNSNVGRSIOCEU-LURJTMIESA-N
Synonym s-+-glycidyl butyrate, s-glycidyl butyrate, s-oxiran-2-ylmethyl butyrate, 2s-oxiran-2-yl methyl butanoate, butanoic acid, 2s-oxiranylmethyl ester, s-glycidyl butanoate, pubchem5837, s-2-butyryloxymethyl oxirane, butyric acid s-glycidyl ester
PubChem CID 6999819
IUPAC Name [(2S)-oxiran-2-yl]methyl butanoate
SMILES CCCC(=O)OCC1CO1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Glycidyl Butyrate