accessibility menu, dialog, popup

UserName

CAS RN 65031-96-1

IUPAC Name: (oxiran-2-yl)methyl butanoate
Synonyms: oxiran-2-ylmethyl butanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: YLNSNVGRSIOCEU-UHFFFAOYNA-N
SMILES: CCCC(=O)OCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results