missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl 3-Nitrobenzenesulfonate 98.0+%, TCI America™

Supplier:  TCI America G028725G

 View more versions of this product

Catalog No. G028725G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-Glycidyl 3-Nitrobenzenesulfonate
65°C
C9H9NO6S
25 g
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
259.23
259.23
Crystalline Powder
115314-14-2
Yellow
MFCD00064582
1325
AIHIHVZYAAMDPM-QMMMGPOBSA-N
[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
146490
≥98.0% (GC)

Chemical Identifiers

115314-14-2
259.23
AIHIHVZYAAMDPM-QMMMGPOBSA-N
146490
[O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
C9H9NO6S
MFCD00064582
s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
[(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DB7192010

TSCA : No

SDS
Documents

Documents

Promotions

Promotions