missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl 3-Nitrobenzenesulfonate 98.0+%, TCI America™

Catalog No. G02875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02875G 5 g
G028725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02875G Supplier TCI America Supplier No. G02875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115314-14-2
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.23
MDL Number MFCD00064582
InChI Key AIHIHVZYAAMDPM-QMMMGPOBSA-N
Synonym s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
PubChem CID 146490
IUPAC Name [(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
SMILES [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1

Specifications

Melting Point 65°C
Color Yellow
Quantity 5 g
UN Number 1325
Formula Weight 259.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-Glycidyl 3-Nitrobenzenesulfonate
RTECSNumber DB7192010
TSCA No