Learn More
(S)-(-)-Glycidol, 99+%, ee 99+%
CAS: 60456-23-7 | C3H6O2 | 74.079 g/mol
Supplier: Thermo Scientific Chemicals H6350206
Description
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
| (S)-(-)-Glycidol | |
| 60456-23-7 | |
| 1.116 g/mL | |
| 66°C (151°F) | |
| 1.433 | |
| 5 g | |
| 79783 | |
| Soluble in chloroform,DMSO and methanol. | |
| C1C(O1)CO | |
| 74.079 | |
| CHEBI:38690 | |
| ≥99%, ee |
| ee ≥99% | |
| Yellow | |
| 66°C to 67°C (19 mmHg) | |
| C3H6O2 | |
| MFCD00074874 | |
| UN2810 | |
| s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s | |
| CTKINSOISVBQLD-VKHMYHEASA-N | |
| [(2S)-oxiran-2-yl]methanol | |
| 6973630 | |
| 74.08 | |
| Liquid |
Chemical Identifiers
| 60456-23-7 | |
| 74.079 | |
| CTKINSOISVBQLD-VKHMYHEASA-N | |
| 6973630 | |
| [(2S)-oxiran-2-yl]methanol |
| C3H6O2 | |
| MFCD00074874 | |
| s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s | |
| CHEBI:38690 | |
| C1C(O1)CO |
Safety and Handling
GHS H Statement
H331-H350-H360-H341-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H331-H335-H340-H350-H360
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber : RR0508100
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only