missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Glycidol, 99+%, ee 99+%
Description
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Color | Yellow |
| Density | 1.116 g/mL |
| Boiling Point | 66°C to 67°C (19 mmHg) |
| Flash Point | 66°C (151°F) |
| Refractive Index | 1.433 |
| Quantity | 1 g |
| UN Number | UN2810 |
| Beilstein | 79783 |
| Solubility Information | Soluble in chloroform,DMSO and methanol. |
| Formula Weight | 74.08 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312-H315-H319-H331-H335-H340-H350-H360 |
| Hazard Statement | GHS H Statement H331-H350-H360-H341-H227-H302-H312-H315-H319-H335 Toxic if inhaled. May cause cancer. May damage fertility or the unborn child. Suspected of causing genetic defects. Combustible liquid. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. |
| Precautionary Statement | P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| RTECSNumber | RR0508100 |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only