missing translation for 'onlineSavingsMsg'
Learn More

S-Furfuryl Thiopropionate 98.0+%, TCI America™

Catalog No. F057825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F057825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F057825G Supplier TCI America Supplier No. F057825G
Only null left

Chemical Identifiers

CAS 59020-85-8
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00040264
InChI Key JNVPDFNCAUOOIT-UHFFFAOYSA-N
Synonym s-furfuryl thiopropionate, propanethioic acid, s-2-furanylmethyl ester, s-furfuryl propanethioate, furfurylthiol propionate, s-2-furylmethyl propanethioate, s-2-furanylmethyl propanethioate, s-furan-2-ylmethyl propanethioate, unii-u05mdn4a54, 1-furan-2-ylmethyl sulfanyl propan-1-one, 1-2-furylmethylthio propan-1-one
PubChem CID 62143
IUPAC Name 1-{[(furan-2-yl)methyl]sulfanyl}propan-1-one
SMILES CCC(=O)SCC1=CC=CO1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 170.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Furfuryl Thiopropionate