accessibility menu, dialog, popup

UserName

H3COSO

CAS RN 59020-85-8

IUPAC Name: 1-{[(furan-2-yl)methyl]sulfanyl}propan-1-one
Synonyms: 1-[(furan-2-ylmethyl)sulfanyl]propan-1-one
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: JNVPDFNCAUOOIT-UHFFFAOYSA-N
SMILES: CCC(=O)SCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results