missing translation for 'onlineSavingsMsg'
Learn More

s-Diphenylcarbazone, ACS Grade, LabChem™

Catalog No. LC136757
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC136757 10 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC136757 Supplier LabChem Supplier No. LC136757

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 538-62-5 , 140-22-7
Molecular Formula C13H12N4O
Molecular Weight (g/mol) 240.266
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
PubChem CID 10860
IUPAC Name 1-anilino-3-phenyliminourea
SMILES C1=CC=C(C=C1)NNC(=O)N=NC2=CC=CC=C2

Specifications

CAS Max % 51, 49
Color Orange
Packaging Amber Glass
Linear Formula C13H14N4O
Quantity 10 g
UN Number UN1325
Sensitivity Passes Test
Solubility Information Soluble in water
Formula Weight 242.28
Grade ACS
Physical Form Powder
Chemical Name or Material s-Diphenylcarbazone
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature