missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol, 99+%

Catalog No. AC290420010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290420010 1 g Glass bottle
AC290420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290420010 Supplier Thermo Scientific Chemicals Supplier No. 290420010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112068-01-6 | C17H19NO | 253.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112068-01-6
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.34
MDL Number MFCD00075506
InChI Key OGCGXUGBDJGFFY-INIZCTEOSA-N
Synonym s---alpha,alpha-diphenyl-2-pyrrolidinemethanol, s-diphenyl pyrrolidin-2-yl methanol, s---2-diphenylhydroxymethyl pyrrolidine, alpha,alpha-diphenyl-l-prolinol, diphenyl-l-pyrrolidin-2-yl-methanol, diphenyl-2s-pyrrolidin-2-yl methanol, s-alpha,alpha-diphenyl-2-pyrrolidinemethanol, s---diphenylpyrrolidinemethanol, s---alpha,alpha-diphenyl-2-pyrrolidine methanol, diphenyl 2s-pyrrolidin-2-ylmethanol
PubChem CID 2724899
IUPAC Name diphenyl-[(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 77°C to 80°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 1 g
Fieser 16,254
Specific Rotation Condition −59° (25°C c=3,methanol)
Specific Rotation -59
Formula Weight 253.34
Percent Purity 99+%
Chemical Name or Material (S)-(-)-α, α-Diphenyl-2-pyrrolidinemethanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only