missing translation for 'onlineSavingsMsg'
Learn More

S-Carboxymethyl-L-cysteine, 98%

Catalog No. AC225810050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225810050 5 g Glass bottle
AC225810250 25 g Glass bottle
AC225811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225810050 Supplier Thermo Scientific Chemicals Supplier No. 225810050

CAS: 638-23-3 | C5H9NO4S | 179.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-23-3
Molecular Formula C5H9NO4S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00002614
InChI Key GBFLZEXEOZUWRN-VKHMYHEASA-N
Synonym s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
PubChem CID 193653
ChEBI CHEBI:16163
IUPAC Name (2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
SMILES N[C@@H](CSCC(O)=O)C(O)=O

Specifications

Melting Point 205.0°C to 207.0°C
Color White
Ignition Residue 0.2% max.
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2SCH2CH(NH2)CO2H
Quantity 5 g
Merck Index 15, 1802
Specific Rotation Condition −31° (20°C c=1,10% NaH2CO3)
Specific Rotation − 31.00
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in hot water (3 g/100 mL), acids and alkal, slightly soluble in methanol and acetone
Formula Weight 179.19
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material S-Carboxymethyl-L-cysteine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 211-327-5
RTECSNumber AY4342000
TSCA TSCA

RUO – Research Use Only