missing translation for 'onlineSavingsMsg'
Learn More

S-Butyrylthiocholine iodide, 98%

Catalog No. AAA1262206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1262206 5 g
AAA1262214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1262206 Supplier Thermo Scientific Chemicals Supplier No. A1262206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1866-16-6 | C9H20INOS | 317.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1866-16-6
Molecular Formula C9H20INOS
Molecular Weight (g/mol) 317.229
MDL Number MFCD00011845
InChI Key WEQAAFZDJROSBF-UHFFFAOYSA-M
Synonym butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
PubChem CID 74630
IUPAC Name 2-butanoylsulfanylethyl(trimethyl)azanium;iodide
SMILES CCCC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 172°C to 176°C
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Quantity 5 g
Beilstein 3729509
Sensitivity Hygroscopic; Light sensitive
Formula Weight 317.23
Percent Purity 98%
Chemical Name or Material S-Butyrylthiocholine iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-475-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only