missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Benzoylthiamine O-Monophosphate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 192°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 466.45 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | S-Benzoylthiamine O-Monophosphate |
Safety and Handling
| EINECSNumber | (9)-1318 |
| RTECSNumber | DH6910000 |
| TSCA | No |
| Recommended Storage | Refrigerator |