missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™
$190.13 - $576.00
Chemical Identifiers
| CAS | 26164-26-1 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00064216 |
| InChI Key | DIWVBIXQCNRCFE-QMMMGPOBSA-N |
| Synonym | s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s |
| PubChem CID | 643325 |
| IUPAC Name | (2S)-2-methoxy-2-phenylacetic acid |
| SMILES | COC(C1=CC=CC=C1)C(=O)O |
Chemical Identifiers
| 26164-26-1 | |
| 166.176 | |
| DIWVBIXQCNRCFE-QMMMGPOBSA-N | |
| 643325 | |
| COC(C1=CC=CC=C1)C(=O)O |
| C9H10O3 | |
| MFCD00064216 | |
| s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s | |
| (2S)-2-methoxy-2-phenylacetic acid |
Specifications
| 26164-26-1 | |
| White-Yellow | |
| C9H10O3 | |
| 1 g | |
| DIWVBIXQCNRCFE-QMMMGPOBSA-N | |
| (2S)-2-methoxy-2-phenylacetic acid | |
| 643325 | |
| ≥98.0% (GC,T) | |
| (S)-(+)-alpha-Methoxyphenylacetic Acid |
| 66°C | |
| 130°C | |
| MFCD00064216 | |
| s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s | |
| COC(C1=CC=CC=C1)C(=O)O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No