missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-Methoxyphenylacetic acid, 99%

Catalog No. AAL08748MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08748MD 250 mg
AAL0874803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08748MD Supplier Thermo Scientific Chemicals Supplier No. L08748MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26164-26-1 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26164-26-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00064216
InChI Key DIWVBIXQCNRCFE-QMMMGPOBSA-N
Synonym s-+-alpha-methoxyphenylacetic acid, s-2-methoxy-2-phenylacetic acid, s-methoxyphenylacetic acid, s-alpha-methoxyphenylacetic acid, s-+-methoxyphenylacetic acid, 2s-2-methoxy-2-phenylacetic acid, s-methoxy phenyl acetic acid, s---alpha-methoxyphenylacetic acid, methoxy phenyl acetic acid #, benzeneacetic acid, .alpha.-methoxy-, s
PubChem CID 643325
IUPAC Name (2S)-2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 63°C to 68°C
Boiling Point 164°C to 166°C (18 mmHg)
Quantity 250 mg
Beilstein 2361718
Optical Rotation +146° (c=0.5 in Ethanol)
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-alpha-Methoxyphenylacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 247-492-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only