Learn More
(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%
CAS: 20445-33-4 | C10H8ClF3O2 | 252.617 g/mol
Supplier: Thermo Scientific Chemicals L14332MF
Description
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.
Solubility
Slightly miscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
| (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride | |
| 1.353 | |
| 89°C (192°F) | |
| 1.469 | |
| 50 mg | |
| 3591564 | |
| 14,6280 | |
| Slightly miscible with water. | |
| +137° (c=6.4 in Carbon Tetrachloride) | |
| (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride | |
| 2724611 | |
| ≥98% |
| 20445-33-4 | |
| 213°C to 214°C | |
| C10H8ClF3O2 | |
| MFCD00067105 | |
| UN3265 | |
| Moisture sensitive | |
| s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride | |
| PAORVUMOXXAMPL-SECBINFHSA-N | |
| COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F | |
| 252.617 | |
| 252.62 |
Chemical Identifiers
| 20445-33-4 | |
| 252.617 | |
| PAORVUMOXXAMPL-SECBINFHSA-N | |
| 2724611 | |
| COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F |
| C10H8ClF3O2 | |
| MFCD00067105 | |
| s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride | |
| (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only