missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98+%

CAS: 20445-33-4 | C10H8ClF3O2 | 252.617 g/mol

Supplier:  Thermo Scientific Chemicals L14332MD

 View more versions of this product

Catalog No. AAL14332MD


Only null left
Explore available promotions

Description

Description

(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride is used in the preparation of natural products and pheromones. It is also employed as a derivatization enantiomeric purity reagent used for the determination of enantiomeric purity of alcohol and amines by gas chromatography.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Specifications

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
1.353
89°C (192°F)
1.469
250 mg
3591564
14,6280
Slightly miscible with water.
+137° (c=6.4 in Carbon Tetrachloride)
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
2724611
≥98%
20445-33-4
213°C to 214°C
C10H8ClF3O2
MFCD00067105
UN3265
Moisture sensitive
s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride
PAORVUMOXXAMPL-SECBINFHSA-N
COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F
252.617
252.62

Chemical Identifiers

20445-33-4
252.617
PAORVUMOXXAMPL-SECBINFHSA-N
2724611
COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F
C10H8ClF3O2
MFCD00067105
s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, +-mtpa-cl, unii-472va70ul4, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, +-mtpa chloride, +-mosher's chloride
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only