missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol Trimethylsilyl Ether 98.0+%, TCI America™

Catalog No. p-7127948
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38431G 1 g
D38435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38431G Supplier TCI America Supplier No. D38431G

Chemical Identifiers

CAS 848821-58-9
Molecular Formula C20H27NOSi
Molecular Weight (g/mol) 325.53
MDL Number MFCD08690083
InChI Key RSUHWMSTWSSNOW-UHFFFAOYNA-N
Synonym (S)-(-)-2-[Diphenyl(trimethylsilyloxy)methyl]pyrrolidine, alpha,alpha-Diphenyl-L-prolinol Trimethylsilyl Ether, (S)-Hayashi-Jorgensen Catalyst
PubChem CID 11198150
IUPAC Name 2-{diphenyl[(trimethylsilyl)oxy]methyl}pyrrolidine
SMILES C[Si](C)(C)OC(C1CCCN1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 325.53
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol Trimethylsilyl Ether
TSCA No
Recommended Storage Refrigerator