missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol 98.0+%, TCI America™

Catalog No. p-7127471
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27351G 1 g
D27355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27351G Supplier TCI America Supplier No. D27351G
Only null left

Chemical Identifiers

CAS 112068-01-6
Molecular Formula C17H19NO
Molecular Weight (g/mol) 253.345
MDL Number MFCD00075506
InChI Key OGCGXUGBDJGFFY-INIZCTEOSA-N
Synonym s---alpha,alpha-diphenyl-2-pyrrolidinemethanol, s-diphenyl pyrrolidin-2-yl methanol, s---2-diphenylhydroxymethyl pyrrolidine, alpha,alpha-diphenyl-l-prolinol, diphenyl-l-pyrrolidin-2-yl-methanol, diphenyl-2s-pyrrolidin-2-yl methanol, s-alpha,alpha-diphenyl-2-pyrrolidinemethanol, s---diphenylpyrrolidinemethanol, s---alpha,alpha-diphenyl-2-pyrrolidine methanol, diphenyl 2s-pyrrolidin-2-ylmethanol
PubChem CID 2724899
IUPAC Name diphenyl-[(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 79°C
Color Yellow
Quantity 1 g
Formula Weight 253.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol